Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PK
Artikelnr. MFLX45501-60
Lieferant: Avantor Fluid Handling
CE certificate
Avantor exclusive
Spezifikationen
Alle Produktkonfigurationen anzeigen- Modell:
- 45501-60
- 45508-70
- 45508-76
- 45508-66
- 45501-66
- 45508-60
- 30800-00
- Connection type (port 1):
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Connection type (port 2):
- 10-32 special tapered thread
- 1/4-28 UNF bottom sealing thread
- 1/4-28 UNF thread
- 10-32 UNF thread
- 1/8-27-NPT thread
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- Garantie:
- 3 Months
- 3 Months
- 3 Months
- Derzeitige Ansicht
Spezifikationen
Über diesen Artikel
Leichter Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Diese Adapter biete einen Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde. Außen-Luer- wie Innen-Luer-Verbindungen sind verfügbar. Alle Gewindeverbindungen sind Innengewinde.
- Verbindungstyp:Female Luer to Thread
- Flow pattern:Straight
- Bezeichnung:Innen-Luer x 10-32 UNF-Gewinde, PC, 25/Pck
- Seq:02
- Environmentally Preferable:
- Modell:45501-60
- Connection type (port 1):Female luer
- No. of ports:2
- Garantie:3 Months
- VE:25
- Fitting material:Polycarbonate
- Thread size UNF(M) (port 1):10-32
Frequently Bought Together