pH/ion meter, SevenDirect SD50
pH/Ion meter, SevenDirect SD50 Pure H₂O
Artikelnr. 662-2388
Anbieter: Mettler - Toledo
Springe zu:
Product Details & Documents
Robust bench top meter for accurate pH and ion determinations in the lab and near production.
- Large colour touch screen provides direct measurement and easy data entry
- Instructions guide users through the sensor calibration process
- Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement
Documents
Spezifikationen
- Bezeichnung:pH/Ion meter, SevenDirect SD50 Pure H₂O
- Seq:07
- ISE Genauigkeit:mmol/L, mol/L, mg/L, ppm, pX: ±0.002
- Kalibrierpunkte:1 - 5
- Temperaturgenauigkeit:±0,1 (0...100 °C)
- Temperaturauflösung:0,1 °C
- ISE Auflösung:mmol/L, mol/L, mg/L, ppm: 0,00001 / %: 0,001 / pX: 0,1 / 0,01
- pH - Genauigkeit:±0,002
- pH - Bereich:−2,000...+20,000
- Kalibrierung:Linear, segmented
- Cat. No.:662-2388
- mV - Bereich:−2000,0...+2000,0
- pH - Auflösung:0,1 / 0,01 / 0,001
- mV - Genauigkeit:±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
- mV - Auflösung:1 / 0,1
- ISE Bereich:mmol/L, mol/L, mg/L, ppm: 0,00000 - 999999 / %: 0,000 - 100,000 / pX: −2,000...20,000
- Temperaturbereich:−30,0…+130,0 °C
Accessories
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2416Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solutionEinloggen für Verfügbarkeit1 * 1 ST - 8,10 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2415Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuringEinloggen für Verfügbarkeit1 * 1 ST - 21,30 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2418Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensorEinloggen für Verfügbarkeit1 * 1 ST - 21,30 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2417Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration positionEinloggen für Verfügbarkeit1 * 1 ST - 40,90 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2419Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensorsEinloggen für Verfügbarkeit1 * 1 ST - 25,20 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2421Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode armEinloggen für Verfügbarkeit1 * 1 ST - 202,00 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2420Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensorEinloggen für Verfügbarkeit1 * 1 ST - 25,20 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2422Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vesselsEinloggen für Verfügbarkeit1 * 1 ST - 244,00 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2424Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)Einloggen für Verfügbarkeit1 * 1 ST - 81,20 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2425Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storageEinloggen für Verfügbarkeit1 * 1 ST - 21,30 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2426Plug set, set of country-specific plugs for Power supply 18 W (662-2424)Einloggen für Verfügbarkeit1 * 1 ST - 27,80 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2382Replaceable protective cover for SevenDirect metersEinloggen für Verfügbarkeit1 * 1 ST - 47,30 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2413Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holderEinloggen für Verfügbarkeit1 * 1 ST - 287,00 €
Produktinformationen
- Artikelnr.BezeichnungVerfügbarkeitPreis
- 662-2414Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meterEinloggen für Verfügbarkeit1 * 1 ST - 337,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



