ContactUsLinkComponent
Microscope slides, 76x56 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., (20 x 50), CE marked 1 * 1.000 ST
Artikelnr. MARI1100090093
Lieferant:  MARIENFELD
Microscope slides, 76x56 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., (20 x 50), CE marked 1 * 1.000 ST
Artikelnr. MARI1100090093
Lieferant:  MARIENFELD
Frequently Bought Together