Coupling body plugs
Coupling body plug, with class VI silicone O-ring, MPX series, Polycarbonat
Artikelnr. CPCCMPX30003MB25
Anbieter: CPC COLDER
Springe zu:
Product Details & Documents
Class VI coupling body plug with Class VI silicone O-ring.
Dokumente
Spezifikationen
- Bezeichnung:Coupling body plug, with class VI silicone O-ring, MPX series
- Material:Polycarbonat
- Seq:02