ContactUsLinkComponent
PR-EXK1 Expansion accessories for ATS / ATL pipes - consisting of robust jack, base plate, AT-Clmp special clamp for pulling out the pipes, transport bag. 1 * 1 ST
Artikelnr. IBSA10908F
Lieferant:  IBS Stock Items
PR-EXK1 Expansion accessories for ATS / ATL pipes - consisting of robust jack, base plate, AT-Clmp special clamp for pulling out the pipes, transport bag. 1 * 1 ST
Artikelnr. IBSA10908F
Lieferant:  IBS Stock Items
Frequently Bought Together