Clipboards
Clipboard, A3 landscape, 350×470 mm
Artikelnr. TEKMWW0205/SSC/L
Anbieter: TEKNOMEK
Springe zu:
Product Details & Documents
The clipboards come in a range of options.
- Brushed dull polish finish
- Spring loaded stainless steel clip
- Available in portrait or landscape
- Material: Fully welded 304-grade stainless steel
The stainless steel clipboards come in sizes A4 and A3, and can be supplied with or without a clip at the back to hang them from our hanging rails.
Spezifikationen
- Bezeichnung:Clipboard, A3 landscape
- Seq:05
- B×H:350×470 mm