Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Matrix Plus, CELLvo™

T150, canted, vented, non-pyrogenic, U-shaped, growth area 150 cm², 15 - 22,5 ml, 5
Artikelnr. STBSAF-HPME-T150
Anbieter:   STEMBIOSYS
Matrix Plus, CELLvo™
Artikelnr. STBSAF-HPME-T150
Anbieter:  STEMBIOSYS
CAS Nummer:  
Matrix Plus, CELLvo™
Matrix Plus, CELLvo™

T150, canted, vented, non-pyrogenic, U-shaped, growth area 150 cm², 15 - 22,5 ml, 5

CAS Nummer:  
Preis
572,00 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

CELLvo™ Matrix Plus is a substrate that aims to recapitulate the biochemical, structural and mechanical cues of the native niche by leaving the extracellular matrix intact. Cell-derived matrices create a biologically relevant culture environment for a variety of cell types including pluripotent cells by providing all the cues cells expect to receive in vivo in culture dishes.

  • Matrix made of polystyrene
  • Natural extracellular matrix synthesised in vitro by human perinatal cells
  • Suitable for the isolation and culture of a variety of cell types including pluripotent cells
  • Shown to enhance the differentiation and maturation of stem cells
  • Composed of more than 100 proteins secreted and assembled by human stem cells
  • For research use only

 : CELLvo™ Matrix Plus arrives at room temperature but should be refrigerated at 2 to 8 °C upon arrival to maintain shelf life. Avoid extended exposure to light.

 : CELLvo™ Matrix Plus is produced under aseptic conditions and should be handled using aseptic technique.

 : CELLvo™ Matrix Plus is not for human use. This product should not be used for diagnostic or therapeutic purposes. Handle CELLvo™ related products in accordance with established bio-safety practices.

Spezifikationen

Spezifikationen

  • VE:
    5
  • Bezeichnung:
    T150, canted, vented, non-pyrogenic, U-shaped, growth area 150 cm²
  • Größe:
    15 - 22,5 ml

Spezifikationen