Express Eingabe
Austria
AuftragsverfolgungKontakt
Matrix, CELLvo™ | Avantor

Matrix, CELLvo™

CELLvo™ Matrix 6-well flat bottom plate, growth area 9,5 cm², 1,9 - 2,9 ml, 5
Artikelnr. STBSBM-HPME-6WP
Anbieter:   STEMBIOSYS
Matrix, CELLvo™
Artikelnr. STBSBM-HPME-6WP
Anbieter:  STEMBIOSYS
CAS Nummer:  
Matrix, CELLvo™
Matrix, CELLvo™

CELLvo™ Matrix 6-well flat bottom plate, growth area 9,5 cm², 1,9 - 2,9 ml, 5

CAS Nummer:  
Preis
184,00 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

CELLvo™ Matrix is a ready to use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.

  • Matrix made of polystyrene
  • Mesenchymal stem cells grown on CELLvo™ Matrix proliferate more rapidly
  • Mesenchymal stem cells grown on CELLvo™ Matrix maintain a smaller cell size
  • Mesenchymal stem cells grown on CELLvo™ Matrix exhibit a higher percentage of cells expressing SSEA-4

CELLvo™ Matrix is a natural extracellular matrix synthesised in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow. It is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.

CELLvo™ Matrix is a protein extracellular matrix that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.

Zubehördetails: Accessories not included.

Lieferumfang: CELLvo™ Matrix arrives at room temperature but should be refrigerated at 2 to 8°C upon arrival. Avoid extended exposure to light.

Verpackung: CELLvo™ Matrix is produced under aseptic conditions and should be handled using aseptic technique.

Achtung! Bitte beachten: CELLvo™ Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes.. Handle CELLvo™ related products in accordance with established bio-safety practices.

Spezifikationen

Spezifikationen

  • VE:
    5
  • Bezeichnung:
    CELLvo™ Matrix 6-well flat bottom plate, growth area 9,5 cm²
  • Größe:
    1,9 - 2,9 ml

Spezifikationen

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.