ContactUsLinkComponent
pF set A with ceramic plates Set for pF determination with ceramic plates, minimal standard set, (0.1...15 bars of suction) 1 * 1 SET
Artikelnr. EIJK0825SA
pF set A with ceramic plates Set for pF determination with ceramic plates, minimal standard set, (0.1...15 bars of suction) 1 * 1 SET
Artikelnr. EIJK0825SA
Frequently Bought Together