pF set A with ceramic plates Set for pF determination with ceramic plates, minimal standard set, (0.1...15 bars of suction) 1 * 1 SET
Artikelnr. EIJK0825SA
Lieferant: EIJKELKAMP AGRISECTOR
pF set A with ceramic plates Set for pF determination with ceramic plates, minimal standard set, (0.1...15 bars of suction) 1 * 1 SET
Artikelnr. EIJK0825SA
Lieferant: EIJKELKAMP AGRISECTOR
Frequently Bought Together
Wasser, HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Lieferant: VWR Chemicals
Sealing film for manual application, Parafilm® M
Lieferant: HEATHROW
Aceton ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Lieferant: KIMBERLY CLARK
VWR®, Serologische Pipetten, Standard Serie
Lieferant: VWR Collection
VWR® Gesichtstücher
Lieferant: VWR Collection
VWR®, Nitrilhandschuhe
Lieferant: VWR Collection
pH-Indikatorstäbchen, MQuant®, Supelco®
Lieferant: Merck
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Lieferant: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschuhe
Lieferant: VWR Collection
VWR®, Transferpipetten, High Performance
Lieferant: VWR Collection