ContactUsLinkComponent
Microsope slides, 75x75mm, thickness 1mm pure white glass, cut edges, twin-frosted approx. 20mm, VE=50, wrapped in paper, Umverpackung 1000 Stk. Nr. 1110252 1 * 1.000 ST
Artikelnr. MARI01110252
Lieferant:  MARIENFELD
Microsope slides, 75x75mm, thickness 1mm pure white glass, cut edges, twin-frosted approx. 20mm, VE=50, wrapped in paper, Umverpackung 1000 Stk. Nr. 1110252 1 * 1.000 ST
Artikelnr. MARI01110252
Lieferant:  MARIENFELD
Frequently Bought Together