Express Eingabe
Austria
AuftragsverfolgungKontakt
Human IL-10 ELISPOT Set | Avantor

Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set
Artikelnr. ABCAAB46549-20X96
Anbieter:   Abcam
Human IL-10 ELISPOT Set
Artikelnr. ABCAAB46549-20X96
Anbieter:  Abcam
CAS Nummer:  
Human IL-10 ELISPOT Set
Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set

CAS Nummer:  
Preis
2 790,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents

Human IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-10 production in a single cell suspension.

Spezifikationen

Spezifikationen

  • Bezeichnung:
    Human IL-10 ELISPOT Set
  • Cat. No.:
    ABCAAB46549-20X96
  • Lagertemperatur:
    4 °C
  • Form:
    Uncoated
  • Species reactivity:
    Human
  • Probentyp:
    Suspension cells
  • Processing time:
    Multiple steps
  • Antigen-Symbol:
    IL-10
  • Regulatory status:
    RUO

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.