AuftragsverfolgungKontakt
Vectors, pBlueScript II | Avantor
Vectors, pBlueScript II
pBluescript II KS(+) Phagemid Kit, f1 origin in (+) orientation, Kpn-Sac polylinker orientation, contains: 20 μg pBluescript II KS(+) phagemid vector, host strain: XL1-Blue MRF', 1 kt
Artikelnr. AGLS212207
Anbieter:   AGILENT
Vectors, pBlueScript II
Artikelnr. AGLS212207
Anbieter:  AGILENT
CAS Nummer:  
Vectors, pBlueScript II
pBluescript II KS(+) Phagemid Kit, f1 origin in (+) orientation, Kpn-Sac polylinker orientation, contains: 20 μg pBluescript II KS(+) phagemid vector, host strain: XL1-Blue MRF', 1 kt
CAS Nummer:  
Preis
642,00 €
Einloggen für Verfügbarkeit
1 * 1 kt
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Spezifikationen
Spezifikationen
  • VE:
    1 kt
  • Bezeichnung:
    pBluescript II KS(+) Phagemid Kit, f1 origin in (+) orientation, Kpn-Sac polylinker orientation, contains: 20 μg pBluescript II KS(+) phagemid vector, host strain: XL1-Blue MRF'