ContactUsLinkComponent
REPROSILPUR ODS-3 GUARD COLUMN 5 X 4.6MM 1 * 5 ST
Artikelnr. MAISR15.93.V0046
Lieferant:  DR MAISCH
REPROSILPUR ODS-3 GUARD COLUMN 5 X 4.6MM 1 * 5 ST
Artikelnr. MAISR15.93.V0046
Lieferant:  DR MAISCH
Frequently Bought Together