ContactUsLinkComponent
Bi-partite gouge auger set, to 5 m, bay Bi-partite gouge auger set. Standard set for sampling in more or less soft soils to a depth of 5 m (bayonet connection), with carrying bag. 1 * 1 SET
  EIJK0402SA
Bi-partite gouge auger set, to 5 m, bay Bi-partite gouge auger set. Standard set for sampling in more or less soft soils to a depth of 5 m (bayonet connection), with carrying bag. 1 * 1 SET
  EIJK0402SA
Frequently Bought Together