Express Eingabe
Austria
AuftragsverfolgungKontakt
COLOUR REFERENCE SOLUTION EU PHARMAC. Y7 1 * 100 mL | Avantor

COLOUR REFERENCE SOLUTION EU PHARMAC. Y7 1 * 100 mL

Artikelnr. REGEEPY701
Anbieter:   REAGECON
COLOUR REFERENCE SOLUTION EU PHARMAC. Y7 1 * 100 mL
Artikelnr. REGEEPY701
Anbieter:  REAGECON
CAS Nummer:  
COLOUR REFERENCE SOLUTION EU PHARMAC. Y7 1 * 100 mL
CAS Nummer:  
Preis
148,00 €
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.