Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Polystyrene centrifuge tubes, PerformR®

PerformR® polystyrene centrifuge tubes, 15 ml, 118 mm, Klar, 3000×g, Flat screw cap, non sterile, Bulk
  3115-870-028-9
 :   Labcon
Polystyrene centrifuge tubes, PerformR®
  3115-870-028-9
 :  Labcon
Polystyrene centrifuge tubes, PerformR®

PerformR® polystyrene centrifuge tubes, 15 ml, 118 mm, Klar, 3000×g, Flat screw cap, non sterile, Bulk

 
418,00 €
   
1 * 500 ST
 

Polysterene tubes are made of crystal clear resin and have bold etched graduations. They can be used for some culture applications that require a glass clear material.

  • ATP-, RNase/Dnase-, and endotoxin-free (non-pyrogenic)
  • Attached flat screw caps with a polyethelene liner
  • Molded graduations and writing surface
  • Polystyrene USP Class VI resin, free of DiHEMDA, oleamide, and BPA
  • Not freezable, not autoclavable

Caps feature a three-ply, co-extruded liner consisting of a foamed white low density polyethelene (LDPE) core sandwiched between two solid layers of LDPE, which contains high density polyethelene (HDPE) for improved stiffness.

 

 
  • Verpackt:
    Bulk
  • Typ:
    Flat screw cap, non sterile
  • Kapazität:
    15 ml
  • Farbe:
    Klar
  • Bezeichnung:
    PerformR® polystyrene centrifuge tubes
  • Länge:
    118 mm
  • Seq:
    01
  • RZB max.:
    3000×g