Barium AAS Standards, (CRMs), Fluka™
Barium, Ba, 10000 mg/L, 5% HCl, Plastic bottle, 100 mL
Catalog # HONCE4106-100ML
Supplier: Honeywell Chemicals
Barium AAS Standards, (CRMs), Fluka™
Catalog # HONCE4106-100ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Konzentration:10000 mg/L
- Matrix:5% HCl
- Packung:Plastic bottle
- VE:100 mL
- Element:Barium
- Symbol:Ba



