Barium AAS Standards, (CRMs), Fluka™
Barium, Ba, 1000 mg/L, 2% HNO₃, Plastic bottle, 100 mL
Artikelnr. HONCE4008-100ML
Anbieter: Honeywell Chemicals
Barium AAS Standards, (CRMs), Fluka™
Artikelnr. HONCE4008-100ML
Anbieter: Honeywell Chemicals
CAS Nummer:
Springe zu:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Spezifikationen
Spezifikationen
- Konzentration:1000 mg/L
- Matrix:2% HNO₃
- Packung:Plastic bottle
- VE:100 mL
- Element:Barium
- Symbol:Ba



