ContactUsLinkComponent
Antibody-Alexa Fluor® 555 Concentration,,,, 1 mg/ml Reactive Species,, Human,Mouse,Rat Host Species,,,, Rabbit Immunogen Synthesized peptide derived from Collagen I . at AA range: 1200-1217 Specificity,,,, Collagen I Polyclonal Antibody detects endog 1 * 100 µG
Artikelnr. GENOITT5769-100U-5
Lieferant:  G-Biosciences
Antibody-Alexa Fluor® 555 Concentration,,,, 1 mg/ml Reactive Species,, Human,Mouse,Rat Host Species,,,, Rabbit Immunogen Synthesized peptide derived from Collagen I . at AA range: 1200-1217 Specificity,,,, Collagen I Polyclonal Antibody detects endog 1 * 100 µG
Artikelnr. GENOITT5769-100U-5
Lieferant:  G-Biosciences
Frequently Bought Together