Objectives
HI PLAN phase objective PH1, 10×/0,25, 12 mm
Catalog # LEIM11506230
Supplier: LEICA MICROSYSTEMS
Jump to:
Product Details & Documents
Plan achromatic and phase contrast objectives.
Specifications
- Vergrößerung:10×/0,25
- Bezeichnung:HI PLAN phase objective PH1
- Arbeitsabstand:12 mm