WP-4C Water Potential Meter Precise and fast system for water-potential measurements (-0.1 MPa to -300 MPa range). With adjustable, internal temperature control. For soil samples using chilled mirror method. Incl. power cord, 10 stainless steel cups 1 * 1 ST
Artikelnr. IBSA40415
Lieferant: IBS Stock Items
Frequently Bought Together