Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Ziprasidone Related Compounds, Reference Standard

Ziprasidone Related Compound F, USP reference standard, Vial, 15 mg
Artikelnr. 1724474.
Anbieter:   USP
Ziprasidone Related Compounds, Reference Standard
Artikelnr. 1724474.
Anbieter:  USP
CAS Nummer:  
Ziprasidone Related Compounds, Reference Standard

Ziprasidone Related Compound F, USP reference standard, Vial, 15 mg

CAS Nummer:  
Preis
1 180,00 €
Einloggen für Verfügbarkeit
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Spezifikationen

Spezifikationen

  • Packung:
    Vial
  • VE:
    15 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Ziprasidone Related Compound F
  • Cat. No.:
    1724474.
  • Synonym(s):
    Sodium 2-(2-amino-5-{2-[4-(benzo[d]isothiazol-3-yl)piperazin-1-yl]ethyl}-4-chlorophenyl)acetate monohydrate
  • Bezeichnung:
    Ziprasidone Related Compound F

Spezifikationen