ContactUsLinkComponent
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP0055 48617.1). 1 * 100 µG
Catalog # ACRBB72-C52HB100UG
Supplier:  ACRO Biosystems
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP0055 48617.1). 1 * 100 µG
Catalog # ACRBB72-C52HB100UG
Supplier:  ACRO Biosystems
Frequently Bought Together