ContactUsLinkComponent
Human IL-15, Tag Free (SPR verified) (IL5-H4117) is expressed from E.coli cells. It contains AA Asn 49 - Ser 162 (Accession P40933- 1). 1 * 50 µG
Artikelnr. ACRBIL5-H4117-50UG
Lieferant:  ACRO Biosystems
Human IL-15, Tag Free (SPR verified) (IL5-H4117) is expressed from E.coli cells. It contains AA Asn 49 - Ser 162 (Accession P40933- 1). 1 * 50 µG
Artikelnr. ACRBIL5-H4117-50UG
Lieferant:  ACRO Biosystems
Frequently Bought Together