ContactUsLinkComponent
ActiveMax® Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P13232). 1 * 50 µG
Artikelnr. ACRBIL7-H4219-50UG
Lieferant:  ACROBIOSYSTEMS
ActiveMax® Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P13232). 1 * 50 µG
Artikelnr. ACRBIL7-H4219-50UG
Lieferant:  ACROBIOSYSTEMS
Frequently Bought Together