Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP00 0409.1). 1 * 1 mg
Catalog # ACRBILR-H5221-1MG
Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP00 0409.1). 1 * 1 mg
Catalog # ACRBILR-H5221-1MG
Frequently Bought Together