Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C 1 * 100 µG
Artikelnr. ACRBNE4-C52H4100UG
Lieferant: ACRO Biosystems
Frequently Bought Together