ContactUsLinkComponent
Human ICAM-1, Fc Tag (IC1-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession AAH15969). 1 * 200 µG
Artikelnr. ACRBIC1-H5250200UG
Lieferant:  ACROBIOSYSTEMS
Human ICAM-1, Fc Tag (IC1-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession AAH15969). 1 * 200 µG
Artikelnr. ACRBIC1-H5250200UG
Lieferant:  ACROBIOSYSTEMS
Frequently Bought Together