ContactUsLinkComponent
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 25 µG
Catalog # ACRBEGR-HF2H5-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 25 µG
Catalog # ACRBEGR-HF2H5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together