Human CD40, Fc Tag (HPLC-verified) (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accessio n P25942-1). 1 * 100 µG
Human CD40, Fc Tag (HPLC-verified) (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accessio n P25942-1). 1 * 100 µG