ContactUsLinkComponent
Human CD47, Fc Tag (HPLC-verified) (CD7-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessio n NP942088). 1 * 100 µG
Artikelnr. ACRBCD7-H5256100UG
Lieferant:  ACRO Biosystems
Human CD47, Fc Tag (HPLC-verified) (CD7-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessio n NP942088). 1 * 100 µG
Artikelnr. ACRBCD7-H5256100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together