Human CD47, Mouse IgG2a Fc Tag, low endotoxin (CD7-H52A5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 13 9 (Accession NP942088). 1 * 100 µG
Artikelnr. ACRBCD7-H52A5100UG
Lieferant: ACROBIOSYSTEMS
Frequently Bought Together