Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630 (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg
Artikelnr. ACRBCD8-H5247-1MG
Lieferant: ACROBIOSYSTEMS
Frequently Bought Together