ContactUsLinkComponent
Recombinat Protein,Human E-Cadherin Cadherin-1 Protein, Fc Tag,Human E-Cadherin, Fc Tag (ECD-H5250) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Pro 621 (Accession AAI41839).This protein carries a human IgG1 Fc tag at the C-t 1 * 100 µG
Artikelnr. ACRBECD-H5250100UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human E-Cadherin Cadherin-1 Protein, Fc Tag,Human E-Cadherin, Fc Tag (ECD-H5250) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Pro 621 (Accession AAI41839).This protein carries a human IgG1 Fc tag at the C-t 1 * 100 µG
Artikelnr. ACRBECD-H5250100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together