ContactUsLinkComponent
Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 200 µG
Artikelnr. ACRBEA3-H5258200UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 200 µG
Artikelnr. ACRBEA3-H5258200UG
Lieferant:  ACRO Biosystems
Frequently Bought Together