ContactUsLinkComponent
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 20 µG
Artikelnr. ACRBGMF-H4214-20UG
Lieferant:  ACROBIOSYSTEMS
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 20 µG
Artikelnr. ACRBGMF-H4214-20UG
Lieferant:  ACROBIOSYSTEMS
Frequently Bought Together