ContactUsLinkComponent
ActiveMax® Human IL-6, Tag Free (IL6-H4218) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Met 212 (Accession NP000591). 1 * 50 µG
Artikelnr. ACRBIL6-H4218-50UG
Lieferant:  ACROBIOSYSTEMS
ActiveMax® Human IL-6, Tag Free (IL6-H4218) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Met 212 (Accession NP000591). 1 * 50 µG
Artikelnr. ACRBIL6-H4218-50UG
Lieferant:  ACROBIOSYSTEMS
Frequently Bought Together