ActiveMax® Human IL-6, Tag Free (IL6-H4218) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Met 212 (Accession NP000591). 1 * 20 µG
ActiveMax® Human IL-6, Tag Free (IL6-H4218) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Met 212 (Accession NP000591). 1 * 20 µG