ContactUsLinkComponent
Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession NP001973. 2). 1 * 1 mg
Catalog # ACRBER3-H5288-1MG
Supplier:  ACRO Biosystems
Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession NP001973. 2). 1 * 1 mg
Catalog # ACRBER3-H5288-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together