ContactUsLinkComponent
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1) . 1 * 100 µG
Artikelnr. ACRBER4-H5221100UG
Lieferant:  ACRO Biosystems
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1) . 1 * 100 µG
Artikelnr. ACRBER4-H5221100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together