ContactUsLinkComponent
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 1 mg
Artikelnr. ACRBCDE-H5222-1MG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 1 mg
Artikelnr. ACRBCDE-H5222-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together