ContactUsLinkComponent
Biotinylated Human OX40, Fc,Avitag (OX0-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accessio n NP003318). 1 * 25 µG
Artikelnr. ACRBOX0-H82F7-25UG
Lieferant:  ACRO Biosystems
Biotinylated Human OX40, Fc,Avitag (OX0-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accessio n NP003318). 1 * 25 µG
Artikelnr. ACRBOX0-H82F7-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together