Biotinylated Human OX40, Fc,Avitag (OX0-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accessio n NP003318). 1 * 25 µG
Biotinylated Human OX40, Fc,Avitag (OX0-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accessio n NP003318). 1 * 25 µG