ContactUsLinkComponent
Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It con tains AA Leu 20 - Pro 219 (Accession AAI13679). 1 * 200 µG
Catalog # ACRBPD2-H82E8200UG
Supplier:  ACRO Biosystems
Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It con tains AA Leu 20 - Pro 219 (Accession AAI13679). 1 * 200 µG
Catalog # ACRBPD2-H82E8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together