ContactUsLinkComponent
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag™,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhis 1 * 200 µG
Artikelnr. ACRBLA1-H82E3200UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag™,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is express ed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhis 1 * 200 µG
Artikelnr. ACRBLA1-H82E3200UG
Lieferant:  ACRO Biosystems
Frequently Bought Together