Biotinylated Human B7-H3, Avitag,His Tag (B73-H82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Ac cession Q5ZPR3-2). 1 * 25 µG
Artikelnr. ACRBB73-H82E6-25UG
Lieferant: ACRO Biosystems
Frequently Bought Together