ContactUsLinkComponent
Biotinylated Human Nectin-2, Fc,Avitag (CD2-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Acce ssion Q92692-2). 1 * 200 µG
Catalog # ACRBCD2-H82F8200UG
Supplier:  ACRO Biosystems
Biotinylated Human Nectin-2, Fc,Avitag (CD2-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Acce ssion Q92692-2). 1 * 200 µG
Catalog # ACRBCD2-H82F8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together