Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (B71-H52A4) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 24 2 (Accession P33681-1). 1 * 1 mg
Artikelnr. ACRBB71-H52A4-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together