Biotinylated Human IL-6 R alpha, Avitag,His Tag (CD6-H82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession P08887-1). 1 * 25 µG
Catalog # ACRBCD6-H82E8-25UG
Supplier: ACRO Biosystems
Frequently Bought Together