ContactUsLinkComponent
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 100 µG
Artikelnr. ACRBAN3-H5226100UG
Lieferant:  ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 100 µG
Artikelnr. ACRBAN3-H5226100UG
Lieferant:  ACROBIOSYSTEMS
Frequently Bought Together