ContactUsLinkComponent
Human BTLA, Fc Tag (BTA-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1). 1 * 1 mg
Artikelnr. ACRBBTA-H5256-1MG
Lieferant:  ACRO Biosystems
Human BTLA, Fc Tag (BTA-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1). 1 * 1 mg
Artikelnr. ACRBBTA-H5256-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together